OriGene
Manufacturer Code:TA804471
Catalog # 50-168-0264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5B3 |
Immunogen | Full length human recombit protein of human PAFAH1B3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PAF acetylhydrolase 29 kDa subunit; PAF-AH 29 kDa subunit; PAF-AH subunit gamma; PAF-AH1b alpha 1 subunit; PAFAH subunit gamma; PAFAHG; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa); Platelet-activating factor acetylhydrolase IB subunit alpha1; platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa)
Gene Aliases: PAFAH1B3; PAFAHG
UniProt ID: (Human) Q15102
Entrez Gene ID: (Human) 5050
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.