Invitrogen
Manufacturer Code:PA531152
Catalog # PIPA531152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 229 of Human PAFAH1B2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 303; intracellular platelet-activating factor acetylhydrolase alpha 2 subunit; PAF acetylhydrolase 30 kDa subunit; PAF acetylhydrolase 30 kDa subunit PAF-AH 30 kDa subunit PAF-AH subunit beta PAF-AH1b alpha 2 subunit PAFAH subunit beta intracellular platelet-activating factor acetylhydrolase alpha 2 subunit platelet-activating factor acetylhydrolase IB subunit beta pl; PAF-AH 30 kDa subunit; PAF-AH subunit beta; PAF-AH1b alpha 2 subunit; PAFAH subunit beta; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa); Platelet-activating factor acetylhydrolase IB subunit alpha2; platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform 1b, alpha2 subunit; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa)
Gene Aliases: AI747451; AU021353; HEL-S-303; mus[b]; PAFAH1B2; PAFAHB
UniProt ID: (Human) P68402, (Mouse) Q61206
Entrez Gene ID: (Human) 5049, (Mouse) 18475
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.