Invitrogen
Manufacturer Code:PA129511
Catalog # PIPA129511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 1-229 of PAFAH1B2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 303; intracellular platelet-activating factor acetylhydrolase alpha 2 subunit; PAF acetylhydrolase 30 kDa subunit; PAF-AH 30 kDa subunit; PAF-AH subunit beta; PAF-AH1b alpha 2 subunit; PAFAH subunit beta; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa); Platelet-activating factor acetylhydrolase IB subunit alpha2; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa)
Gene Aliases: HEL-S-303; PAFAH1B2; PAFAHB
UniProt ID: (Human) P68402
Entrez Gene ID: (Human) 5049
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.