Novus Biologicals
Manufacturer Code:NBP219677
Catalog # NBP219677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PAFAH1B2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1.47 intracellular platelet-activating factor acetylhydrolase alpha 2 subunit PAF acetylhydrolase 30 kDa subunit PAF-AH 30 kDa subunit PAFAH subunit beta PAF-AH subunit beta PAF-AH1b alpha 2 subunit PAFAHB platelet-activating factor acetylhydrolase 1b catalytic subunit 2 (30kDa) platelet-activating factor acetylhydrolase IB subunit beta platelet-activating factor acetylhydrolase isoform Ib beta subunit 30kDa platelet-activating factor acetylhydrolase isoform Ib subunit 2 (30kDa); epididymis secretory protein Li 303; intracellular platelet-activating factor acetylhydrolase alpha 2 subunit; PAF acetylhydrolase 30 kDa subunit; PAF-AH 30 kDa subunit; PAF-AH subunit beta; PAF-AH1b alpha 2 subunit; PAFAH subunit beta; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa); Platelet-activating factor acetylhydrolase IB subunit alpha2; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa)
Gene Aliases: HEL-S-303; PAFAH1B2; PAFAHB
UniProt ID: (Human) P68402
Entrez Gene ID: (Human) 5049
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.