Proteintech
Manufacturer Code:147291AP150UL
Catalog # 50-172-9678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PAFAH1B2 Fusion Protein Ag6489 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 303; intracellular platelet-activating factor acetylhydrolase alpha 2 subunit; PAF acetylhydrolase 30 kDa subunit; PAF AH 30 kDa subunit PAF AH subunit beta PAFAH subunit beta PAFAH1B2 PAFAHB; PAF-AH 30 kDa subunit; PAF-AH alpha 2; PAF-AH subunit beta; PAF-AH1b alpha 2 subunit; PAFAH subunit beta; platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa); Platelet-activating factor acetylhydrolase alpha 2 subunit; platelet-activating factor acetylhydrolase alpha 2 subunit (PAF-AH alpha 2); Platelet-activating factor acetylhydrolase IB subunit alpha2; platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform 1b, alpha2 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 2; platelet-activating factor acetylhydrolase, isoform Ib, beta subunit; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2; platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa)
Gene Aliases: AI747451; AU021353; HEL-S-303; mus[b]; PAFAH1B2; PAFAHB
UniProt ID: (Human) P68402, (Rat) O35264, (Mouse) Q61206
Entrez Gene ID: (Human) 5049, (Rat) 64189, (Mouse) 18475
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.