Invitrogen
Manufacturer Code:PA534613
Catalog # PIPA534613
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 100 - 200 of Human PAF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F23149&lowbar1 PD2 RNA polymerase II-associated factor 1 homolog pancreatic differentiation protein 2; hPAF1; Paf1, RNA polymerase II associated factor, homolog; Pancreatic differentiation protein 2; RNA polymerase II-associated factor 1 homolog
Gene Aliases: 5730511K23Rik; AA675045; F23149_1; PAF1; PD2
UniProt ID: (Human) Q8N7H5, (Mouse) Q8K2T8
Entrez Gene ID: (Human) 54623, (Mouse) 54624
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.