Novus Biologicals
Manufacturer Code:NBP246310
Catalog # NBP246310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5C10 |
Immunogen | Human recombinant protein fragment correspongding to amino acids 299-588 of human PADI4(NP_036519) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HL-60 PAD; HL-60 PAD PADI5PADI-H protein PADPAD4 PDI4 PDI5EC 3.5.3.15 peptidyl arginine deiminase type IV peptidyl arginine deiminase type V Peptidylarginine deiminase IV Protein-arginine deiminase type IV protein-arginine deiminase type-4; PADI-H protein; peptidyl arginine deiminase, type IV; peptidyl arginine deiminase, type V; Peptidylarginine deiminase IV; Protein-arginine deiminase type IV; Protein-arginine deiminase type-4
Gene Aliases: PAD; PAD4; PADI4; PADI5; PDI4; PDI5
UniProt ID: (Human) Q9UM07
Entrez Gene ID: (Human) 23569
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.