Proteintech
Manufacturer Code:173731AP150UL
Catalog # 50-100-2667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PADI4 Fusion Protein Ag11068 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HL 60 PAD PAD PAD4 PADI4 PADI5 PDI4 PDI5 Peptidylarginine deiminase IV; HL-60 PAD; PAD type IV; PAD-R4; PADI-H protein; peptidyl arginine deiminase, type 4; peptidyl arginine deiminase, type IV; peptidyl arginine deiminase, type V; Peptidylarginine deiminase IV; Peptidylarginine deiminase type alpha; Protein-arginine deiminase type IV; Protein-arginine deiminase type-4
Gene Aliases: PAD; PAD4; PADI4; PADI5; PDI4; PDI5
UniProt ID: (Human) Q9UM07, (Rat) O88807, (Mouse) Q9Z183
Entrez Gene ID: (Human) 23569, (Rat) 29512, (Mouse) 18602
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.