Novus Biologicals
Manufacturer Code:NBP293672100UL
Catalog # NB029207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 26-210 of human PADI2 (NP_031391.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.5.3.15 KIAA0994PDI2PAD2 PAD-H19 peptidlyarginine deiminase type II peptidyl arginine deiminase type II Peptidylarginine deiminase II protein arginine deiminase Protein-arginine deiminase type II protein-arginine deiminase type-2; PAD-H19; peptidyl arginine deiminase, type II; Peptidylarginine deiminase II; Protein-arginine deiminase type II; Protein-arginine deiminase type-2
Gene Aliases: KIAA0994; PAD-H19; PAD2; PADI2; PDI2
UniProt ID: (Human) Q9Y2J8
Entrez Gene ID: (Human) 11240
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.