Novus Biologicals
Manufacturer Code:NBP299019UV
Catalog # NB098661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PACT. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DYT16interferon-inducible double stranded RNA-dependent activator PACTPKR-associating protein X PKR-associated protein X protein kinase interferon-inducible double stranded RNA dependent activator RAXHSD14; interferon-inducible double stranded RNA-dependent protein kinase activator A; Interferon-inducible double-stranded RNA-dependent protein kinase activator A; PKR-associated protein X; PKR-associating protein X; Protein activator of the interferon-induced protein kinase; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; Protein kinase, interferon-inducible double-stranded RNA-dependent activator
Gene Aliases: DYT16; HSD-14; HSD14; PACT; PRKRA; RAX
UniProt ID: (Human) O75569
Entrez Gene ID: (Human) 8575
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.