OriGene
Manufacturer Code:CF502214
Catalog # 50-166-1139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G7 |
Immunogen | Full length human recombit protein of human PACSIN3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Protein kinase C and casein kinase substrate in neurons protein 3; SDPIII; SH3 domain-containing protein 6511; syndapin III
Gene Aliases: PACSIN3; SDPIII
UniProt ID: (Human) Q9UKS6
Entrez Gene ID: (Dog) 475984, (Human) 29763, (Rat) 311187
Molecular Function: actin family cytoskeletal protein cytoskeletal protein membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.