Invitrogen
Manufacturer Code:PA515356
Catalog # PIPA515356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PACSIN3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein kinase C and casein kinase II substrate protein 3; Protein kinase C and casein kinase substrate in neurons protein 3; Protein kinase C and casein kinase substrate in neurons protein 3 SH3 domain-containing protein 6511 Endophilin I; SH3 domain-containing protein 6511; syndapin III
Gene Aliases: 4921507A02Rik; 6330413E15Rik; AW413130; PACSIN3; SDPIII
UniProt ID: (Human) Q9UKS6, (Mouse) Q99JB8
Entrez Gene ID: (Human) 29763, (Mouse) 80708
Molecular Function: actin family cytoskeletal protein cytoskeletal protein membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.