Novus Biologicals
Manufacturer Code:NBP224481SS
Catalog # NBP224481SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 400-450 of human PACSIN2 was used as the immunogen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytoplasmic phosphoprotein PACSIN2; cytoplasmic phosphoprotein PACSIN2 protein kinase C and casein kinase substrate in neurons 2 protein kinase C and casein kinase substrate in neurons protein 2 SDPII syndapin II; Protein kinase C and casein kinase substrate in neurons protein 2; SdpII; syndapin 2; syndapin II; Syndapin-2; Syndapin-II; syndapin2
Gene Aliases: PACSIN2; SDPII
UniProt ID: (Human) Q9UNF0
Entrez Gene ID: (Human) 11252
Molecular Function: actin family cytoskeletal protein cytoskeletal protein membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.