Invitrogen
Manufacturer Code:PA517994
Catalog # PIPA517994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SSSYDEASLAPEET) corresponding to the N-terminus amino acids of PACSIN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: PACSIN1 protein kinase C and casein kinase substrate in neurons 1 KIAA1379 SDPI syndapin I; Protein kinase C and casein kinase substrate in neurons protein 1; syndapin 1; syndapin I; Syndapin-1
Gene Aliases: KIAA1379; PACSIN1; SDPI
UniProt ID: (Human) Q9BY11
Entrez Gene ID: (Human) 29993
Molecular Function: actin family cytoskeletal protein cytoskeletal protein membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.