Novus Biologicals
Manufacturer Code:NBP233756
Catalog # NBP233756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: EKQPKKAEGVALTNATGAVESTSQAGDRGSVSSYDRGQPYATEWSDDESGN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1379SDPI protein kinase C and casein kinase substrate in neurons 1 protein kinase C and casein kinase substrate in neurons protein 1 syndapin I; Protein kinase C and casein kinase substrate in neurons protein 1; syndapin 1; syndapin I; Syndapin-1
Gene Aliases: KIAA1379; PACSIN1; SDPI
UniProt ID: (Human) Q9BY11
Entrez Gene ID: (Human) 29993
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.