Novus Biologicals
Manufacturer Code:NBP299230F
Catalog # NB059250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PACSIN1. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1379SDPI protein kinase C and casein kinase substrate in neurons 1 protein kinase C and casein kinase substrate in neurons protein 1 syndapin I; Protein kinase C and casein kinase substrate in neurons protein 1; syndapin 1; syndapin I; Syndapin-1
Gene Aliases: KIAA1379; PACSIN1; SDPI
UniProt ID: (Human) Q9BY11
Entrez Gene ID: (Human) 29993
Molecular Function: actin family cytoskeletal protein cytoskeletal protein membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.