Invitrogen
Manufacturer Code:PA572866
Catalog # PIPA572866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PACS2 antibody for a 17 amino acid peptide near the center of human PACS2. The immunogen is within amino acids 330 - 380 of PACS2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PACS-2; PACS1-like protein; Phosphofurin acidic cluster sorting protein 2; Phosphofurin acidic cluster sorting protein 2 FLJ25488 KIAA0602 PACS1L PACS-2
Gene Aliases: 6720425G15Rik; AW208651; AW489976; E230011J22; KIAA0602; mKIAA0602; PACS-2; PACS1L; PACS2
UniProt ID: (Human) Q86VP3, (Mouse) Q3V3Q7
Entrez Gene ID: (Human) 23241, (Mouse) 217893, (Rat) 691631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.