Invitrogen
Manufacturer Code:PA3213
Catalog # PIPA3213
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C-terminal of rat PAC1 (503-523) CLSKSSSQLRMSSLPADNLAT coupled to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenylate cyclase activating polypeptide 1 (pituitary) receptor type I; adenylate cyclase activating polypeptide 1 type 1 receptor; PAC1 receptor PACAP type 1a receptor PACAP-R-1; PACAP type I receptor; PACAP type IA receptor; PACAP-R-1; PACAP-R-1A; pituitary adenylate cyclase activating polypeptide 1 receptor (1); pituitary adenylate cyclase activating polypeptide receptor; Pituitary adenylate cyclase-activating polypeptide type I receptor; pituitary adenylate cyclase-activating polypeptide type IA receptor
Gene Aliases: 2900024I10Rik; Adcyap1r1; AI846590; PAC1; PAC1-R; PAC1R; PACAP-R1; PACAP-R1A; PACAP1-R; PACAPR1; PACAPR1A
UniProt ID: (Mouse) P70205, (Rat) P32215
Entrez Gene ID: (Mouse) 11517, (Rat) 24167
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.