Invitrogen
Manufacturer Code:MA537769
Catalog # PIMA537769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1996CT329.45.62 |
Immunogen | recombinant protein from the human region of human ADCYAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADCYAP1 adenylate cyclase activating polypeptide 1 adenylate cyclase activating polypeptide 1 (pituitary) HSPC190 MGC126852 OTTHUMP00000162201 Pacap pacap 38 PACAP-27 PACAP-38 PACAP-related peptide pERp1 pituitary adenylate cyclase activating polypeptide pituitary adenylate cyclase activating polypeptide 1 pituitary adenylate cyclase-activating polypeptide Pituitary adenylate cyclase-activating polypeptide 27 Pituitary adenylate cyclase-activating polypeptide 38 pituitary adenylate cyclase-activating polypeptide precursor (Pacap) prepro-PACAP PRP-48; adenylate cyclase activating polypeptide 1 (pituitary); PACAP; PACAP-27; PACAP-38; PACAP-related peptide; pituitary adenylate cyclase activating polypeptide; Pituitary adenylate cyclase-activating polypeptide; Pituitary adenylate cyclase-activating polypeptide 27; Pituitary adenylate cyclase-activating polypeptide 38; prepro-PACAP; PRP-48
Gene Aliases: ADCYAP1; PACAP
UniProt ID: (Human) P18509
Entrez Gene ID: (Human) 116
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.