Invitrogen
Manufacturer Code:PA547838
Catalog # PIPA547838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse PACAP/ADCYAP1 Pro26-Leu175 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: ADCYAP1 pituitary adenylate cyclase-activating polypeptide; PACAP; PACAP-27; PACAP-38; PACAP-related peptide; Pituitary adenylate cyclase-activating polypeptide; Pituitary adenylate cyclase-activating polypeptide 27; Pituitary adenylate cyclase-activating polypeptide 38; PRP-48
Gene Aliases: Adcyap1; PACAP
UniProt ID: (Mouse) O70176
Entrez Gene ID: (Mouse) 11516
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.