Invitrogen
Manufacturer Code:PA521794
Catalog # PIPA521794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 189 of PACAP (Uniprot IDNo.Q8WU39) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenylate cyclase activating polypeptide 1 (pituitary); HSPC190 PACAP pERp1 HSPC190 caspase-2 binding protein plasma cell-induced ER protein 1 plasma cell-induced resident ER protein plasma cell-induced resident endoplasmic reticulum protein proapoptotic caspase adapter protein proapoptotic caspase adaptor protein; PACAP; PACAP-27; PACAP-38; PACAP-related peptide; pituitary adenylate cyclase activating polypeptide; Pituitary adenylate cyclase-activating polypeptide; Pituitary adenylate cyclase-activating polypeptide 27; Pituitary adenylate cyclase-activating polypeptide 38; prepro-PACAP; PRP-48
Gene Aliases: ADCYAP1; PACAP
UniProt ID: (Human) P18509
Entrez Gene ID: (Human) 116
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.