Novus Biologicals
Manufacturer Code:NB100622410.1ML
Catalog # NB017449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Proprietary 14 amino acid peptide sequence of PACAP conjugated to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: adenylate cyclase activating polypeptide 1 (pituitary); adenylate cyclase activating polypeptide 1 (pituitary) PACAPMGC126852 pituitary adenylate cyclase-activating polypeptide; PACAP; PACAP-27; PACAP-38; PACAP-related peptide; pituitary adenylate cyclase activating polypeptide; Pituitary adenylate cyclase-activating polypeptide; Pituitary adenylate cyclase-activating polypeptide 27; Pituitary adenylate cyclase-activating polypeptide 38; prepro-PACAP; PRP-48
Gene Aliases: ADCYAP1; PACAP
UniProt ID: (Human) P18509
Entrez Gene ID: (Human) 116
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.