Novus Biologicals
Manufacturer Code:NBP297421F
Catalog # NB043226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PAC1R fragment. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenylate cyclase activating polypeptide 1 (pituitary) receptor type 1 adenylate cyclase activating polypeptide 1 (pituitary) receptor type I PACAP type I receptor PACAP-R1 PACAP-R-1 PACAPRI PACAPRPAC1 pituitary adenylate cyclase activating polypeptide 1 receptor type I Hiphop pituitary adenylate cyclase-activating polypeptide type I receptor; adenylate cyclase activating polypeptide 1 (pituitary) receptor type I; PACAP receptor 1; PACAP type I receptor; PACAP-R1; pituitary adenylate cyclase activating polypeptide 1 receptor type I Hiphop; Pituitary adenylate cyclase-activating polypeptide type I receptor
Gene Aliases: ADCYAP1R1; PAC1; PAC1R; PACAPR; PACAPRI
UniProt ID: (Human) P41586
Entrez Gene ID: (Human) 117
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.