Novus Biologicals
Manufacturer Code:NBP157216
Catalog # NBP157216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PABPN1(poly(A) binding protein nuclear 1) The peptide sequence was selected from the N terminal of PABPN1. Peptide sequence MAAAAAAAAAAGAAGGRGSGPGRRRHLVPGAGGEAGEGAPGGAGDYGNGL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nuclear poly(A)-binding protein 1; Nuclear poly(A)-binding protein 1 OPMD PAB2poly(A) binding protein II PABII PABP-2 PABP2polyadenylate-binding protein 2 poly(A) binding protein 2 poly(A) binding protein nuclear 1 Poly(A)-binding protein 2 Poly(A)-binding protein II poly(A)-binding protein nuclear 1 Polyadenylate-binding nuclear protein 1; PABII; PABP-2; poly(A) binding protein 2; poly(A) binding protein II; poly(A) binding protein, nuclear 1; Poly(A)-binding protein II; Polyadenylate-binding nuclear protein 1; Polyadenylate-binding protein 2
Gene Aliases: OPMD; PAB2; PABII; PABP-2; PABP2; PABPN1
UniProt ID: (Human) Q86U42
Entrez Gene ID: (Human) 8106
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.