Abnova
Manufacturer Code:PAB19984
Catalog # 89-121-486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PABPN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PABPN1.
For Research Use Only
Protein Aliases: Nuclear poly(A)-binding protein 1; OPMD; PAB2; PABII; PABP-2; PABP2; poly(A) binding protein 2; poly(A) binding protein II; poly(A) binding protein, nuclear 1; Poly(A)-binding protein II; Polyadenylate-binding nuclear protein 1; Polyadenylate-binding protein 2
Gene Aliases: OPMD; PAB2; PABII; PABP-2; PABP2; PABPN1
UniProt ID: (Human) Q86U42
Entrez Gene ID: (Human) 8106
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.