Invitrogen
Manufacturer Code:PA529883
Catalog # PIPA529883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 356 and 630 of Human PABP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAB1 PABP PABP1 PABPC2 PABPL1 poly(A) binding protein cytoplasmic 2 polyadenylate-binding protein 1; PABP-1; poly A binding protein 1; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 2; poly(A)-binding protein 1; Polyadenylate-binding protein 1
Gene Aliases: ePAB; PAB1; PABP; PABP1; PABPC1; PABPC2; PabpI; PABPL1
UniProt ID: (Human) P11940, (Mouse) P29341
Entrez Gene ID: (Human) 26986, (Mouse) 18458
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.