Novus Biologicals
Manufacturer Code:NBP299378FR
Catalog # NB085065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PABP. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAB1polyadenylate-binding protein 1 PABP PABP-1 PABP1poly(A) binding protein cytoplasmic 2 PABPC2 PABPL1 poly(A) binding protein cytoplasmic 1 Poly(A)-binding protein 1 poly(A)-binding protein cytoplasmic 2; PABP-1; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 2; Polyadenylate-binding protein 1
Gene Aliases: PAB1; PABP; PABP1; PABPC1; PABPC2; PABPL1
UniProt ID: (Human) P11940
Entrez Gene ID: (Human) 26986
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.