Invitrogen
Manufacturer Code:PA576932
Catalog # PIPA576932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PTPRN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 105 kDa islet cell antigen; BEM-3; Brain-enriched membrane-associated protein tyrosine phosphatase; ICA 512; ICA105; ICA512; ICA512-CCF; ICA512-cleaved cytosolic fragment; ICA512-N-terminal fragment; ICA512-NTF; ICA512-TMF; ICA512-transmembrane fragment; islet cell antigen 2; Islet cell antigen 512; Islet cell autoantigen 3; protein tyrosine phosphatase, receptor type, N; protein tyrosine phosphatase, receptor-type, N; protein tyrosine phosphatase-like N; PTP IA-2; PTPLP; R-PTP-N; Receptor-type tyrosine-protein phosphatase-like N; Receptor-type tyrosine-protein phosphatase-like N R-PTP-N Islet cell antigen 512 ICA 512 Islet cell autoantigen 3 PTP IA-2 PTPRN ICA3 ICA512
Gene Aliases: IA-2; IA-2/PTP; IA2; ICA3; ICA512; mIA-A; mKIAA4064; PTPRN; R-PTP-N
UniProt ID: (Human) Q16849, (Rat) Q63259
Entrez Gene ID: (Human) 5798, (Rat) 116660, (Mouse) 19275
Molecular Function: hydrolase phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.