Novus Biologicals
Manufacturer Code:NBP106542
Catalog # NBP106542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 204 and 254 of human PTIP-associated 1 protein using the numbering given in entry NP_078792.1 (GeneID 79447). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 16 open reading frame 53 FLJ22459 GAS glutamate-rich coactivator interacting with SRC1/NCOA1 MGC4606 PA1 PAXIP1-associated protein 1 PTIP-associated 1 protein PTIP-associated protein 1; GAS; glutamate-rich coactivator associated with SRC1; Glutamate-rich coactivator interacting with SRC1; glutamate-rich coactivator interacting with SRC1/NCOA1; PAXIP1 associated glutamate-rich protein 1; PAXIP1-associated glutamate-rich protein 1; PAXIP1-associated protein 1; PTIP-associated 1 protein; PTIP-associated protein 1
Gene Aliases: C16orf53; GAS; PA1; PAGR1
UniProt ID: (Human) Q9BTK6
Entrez Gene ID: (Human) 79447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.