Invitrogen
Manufacturer Code:PA5114115
Catalog # PIPA5114115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PH-4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4933406E20Rik AI853847 BB128974 EGLN4 FLJ20262 HIFPH4 HIF-PH4 HIF-prolyl hydroxylase 4 HPH-4 hypoxia inducible factor prolyl 4 hydroxylase hypoxia-inducible factor prolyl 4-hydroxylase Hypoxia-inducible factor prolyl hydroxylase 4 P4H TM P4H with transmembrane domain P4htm P4H-TM PH 4 Ph4 PH-4 PHD4 proline 4-hydroxylase prolyl 4-hydroxylase transmembrane prolyl 4-hydroxylase transmembrane (endoplasmic reticulum) Prolyl hydroxlase domain-containing 4 RGD1311848 Transmembrane prolyl 4-hydroxylase; HIF-PH4; HIF-prolyl hydroxylase 4; HPH-4; hypoxia inducible factor prolyl 4 hydroxylase; hypoxia-inducible factor prolyl 4-hydroxylase; Hypoxia-inducible factor prolyl hydroxylase 4; P4H with transmembrane domain; P4H-TM; proline 4-hydroxylase; prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum); Prolyl hydroxlase domain-containing 4; Transmembrane prolyl 4-hydroxylase
Gene Aliases: EGLN4; HIFPH4; P4H-TM; P4HTM; PH-4; PH4; PHD4
UniProt ID: (Human) Q9NXG6
Entrez Gene ID: (Human) 54681
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.