Proteintech
Manufacturer Code:666041IG150UL
Catalog # 50-173-7022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A9A4 |
Immunogen | P4HA2 Fusion Protein Ag4714 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 4 PH alpha 2 P4HA2; 4-PH alpha 2; 4-PH alpha-2; C-P4Halpha(II); collagen prolyl 4-hydroxylase alpha(II); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; Procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; Prolyl 4-hydroxylase alpha IIa subunit; Prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II
Gene Aliases: AA407196; C76437; P4HA2; P4hl; UNQ290/PRO330
UniProt ID: (Human) O15460, (Mouse) Q60716
Entrez Gene ID: (Human) 8974, (Mouse) 18452, (Rat) 360526
Molecular Function:
hydroxylase
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.