Invitrogen
Manufacturer Code:PA570557
Catalog # PIPA570557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human P4HA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-PH alpha 2; 4-PH alpha 2 4-PH alpha-2 C-P4Halpha(II) collagen prolyl 4-hydroxylase alpha(II) procollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) alpha polypeptide II procollagen-proline2-oxoglutarate-4-dioxygenase subunit alpha-2 prolyl 4-hydr; 4-PH alpha-2; C-P4Halpha(II); collagen prolyl 4-hydroxylase alpha(II); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; Procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; prolyl 4-hydroxylase A; Prolyl 4-hydroxylase alpha IIa subunit; Prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II
Gene Aliases: AA407196; C76437; P4HA2; P4hl; UNQ290/PRO330; zgc:91948
UniProt ID: (Human) O15460, (Mouse) Q60716
Entrez Gene ID: (Horse) 100072882, (Bovine) 507327, (Human) 8974, (Dog) 474672, (Pig) 100623801, (Guinea pig) 100718564, (Mouse) 18452, (Rat) 360526, (Zebrafish) 373132
Molecular Function: hydroxylase oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.