Invitrogen
Manufacturer Code:PA527761
Catalog # PIPA527761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 86 and 466 of Human P4HA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-PH alpha 2; 4-PH alpha 2 4-PH alpha-2 C-P4Halpha(II) collagen prolyl 4-hydroxylase alpha(II) procollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) alpha polypeptide II procollagen-proline2-oxoglutarate-4-dioxygenase subunit alpha-2 prolyl 4-hydr; 4-PH alpha-2; C-P4Halpha(II); collagen prolyl 4-hydroxylase alpha(II); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; Procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; Prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II
Gene Aliases: P4HA2; UNQ290/PRO330
UniProt ID: (Human) O15460
Entrez Gene ID: (Human) 8974
Molecular Function: hydroxylase oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.