Novus Biologicals
Manufacturer Code:NB11040494
Catalog # NB11040494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human Prolyl 4-Hydroxylase Alpha-2 using the numbering given in entry NP_004190.1 (GeneID 8974 ). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate-4-dioxygenase subunit alpha-2 4-PH alpha 2 4-PH alpha-2 alpha polypeptide II collagen prolyl 4-hydroxylase alpha(II) C-P4Halpha(II) EC 1.14.11.2 Procollagen-proline procollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) prolyl 4-hydroxylase subunit alpha-2 prolyl 4-hydroxylase alpha polypeptide II; 4-PH alpha 2; 4-PH alpha-2; C-P4Halpha(II); collagen prolyl 4-hydroxylase alpha(II); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; Procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; Prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II
Gene Aliases: P4HA2; UNQ290/PRO330
UniProt ID: (Human) O15460
Entrez Gene ID: (Human) 8974
Molecular Function: hydroxylase oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.