Novus Biologicals
Manufacturer Code:NBP184397
Catalog # NBP184397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:FTSIGQMTDLIHTEKDLVTSLKDYIKAEEDKLEQIKKWAEKLDRLTSTATKDPEGFVGHPVNAFKLMKRLNTEWSELENLVLKDMSDGFISNLTIQRQYFPNDEDQVGAAKALLRLQDTYNLDTDTISKGNLPGVKHKSFLTAEDC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) 2-oxoglutarate-4-dioxygenase subunit alpha-1 4-PH alpha-1 alpha polypeptide I EC 1.14.11.2 Procollagen-proline prolyl 4-hydroxylase alpha polypeptide I; 4-PH alpha-1; C-P4Halpha(I); collagen prolyl 4-hydroxylase alpha(I); procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide I; Procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-1; Prolyl 4-hydroxylase subunit alpha-1; prolyl 4-hydroxylase, alpha polypeptide I
Gene Aliases: P4HA; P4HA1
UniProt ID: (Human) P13674
Entrez Gene ID: (Human) 5033
Molecular Function:
hydroxylase
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.