Invitrogen
Manufacturer Code:PA5106424
Catalog # PIPA5106424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human P3H3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC016431 gene rich cluster B Grcb HSU47926 leprecan-like 2 Leprecan-like protein 2 LEPREL2 P3H3 procollagen-proline 3-dioxygenase prolyl 3-hydroxylase 3 Protein B; gene rich cluster, B; leprecan-like 2; Leprecan-like protein 2; procollagen-proline 3-dioxygenase; Prolyl 3-hydroxylase 3; Protein B
Gene Aliases: BC016431; GRCB; HSU47926; LEPREL2; P3H3
UniProt ID: (Human) Q8IVL6, (Mouse) Q8CG70
Entrez Gene ID: (Human) 10536, (Rat) 297595, (Mouse) 14789
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.