Proteintech
Manufacturer Code:160231AP150UL
Catalog # 50-173-0652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | P3H3 Fusion Protein Ag8850 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gene rich cluster, B; GRCB HSU47926 leprecan like 2 Leprecan like protein 2 LEPREL2 P3H3 Prolyl 3 hydroxylase 3 Protein B; leprecan-like 2; Leprecan-like protein 2; procollagen-proline 3-dioxygenase; Prolyl 3-hydroxylase 3; Protein B
Gene Aliases: BC016431; GRCB; HSU47926; LEPREL2; P3H3
UniProt ID: (Human) Q8IVL6, (Mouse) Q8CG70
Entrez Gene ID: (Human) 10536, (Rat) 297595, (Mouse) 14789
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.