Abnova
Manufacturer Code:PAB8974
Catalog # 89-134-061
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus amino acids 361-373 of rat P2ry2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of P2ry2.
For Research Use Only
Protein Aliases: ATP receptor; P2U purinoceptor 1; P2U1; P2Y ATP receptor 2; P2Y purinoceptor 2; P2Y2; Purinergic receptor
Gene Aliases: P2ru1; P2ry2; P2Y2
UniProt ID: (Rat) P41232
Entrez Gene ID: (Rat) 29597
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.