Invitrogen
Manufacturer Code:PA5111893
Catalog # PIPA5111893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 172 - 186 of mouse P2RY6 (2nd extracellular loop) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010204J23Rik MGC15335 P2ry6 P2Y purinoceptor 6 p2y3 and chick p2y3 receptor P2Y6 PP2891 Purinoceptor p2y6; G-coupled nucleotide receptor; P2 purinoceptor; P2Y ATP receptor 6; P2Y purinoceptor 6; P2Y6; P2Y6 receptor; pyrimidinergic receptor P2Y, G-protein coupled, 6
Gene Aliases: 2010204J23Rik; P2RY6; P2Y6; PP2891
UniProt ID: (Human) Q15077, (Rat) Q63371, (Mouse) Q9ERK9
Entrez Gene ID: (Human) 5031, (Rat) 117264, (Mouse) 233571
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.