Invitrogen
Manufacturer Code:MA534766
Catalog # PIMA534766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG37-77 |
Immunogen | Synthetic peptide corresponding to N-terminal human P2Y6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2010204J23Rik MGC15335 P2ry6 P2Y purinoceptor 6 p2y3 and chick p2y3 receptor P2Y6 PP2891 Purinoceptor p2y6; G-coupled nucleotide receptor; P2 purinoceptor; P2Y purinoceptor 6; P2Y6; P2Y6 receptor; pyrimidinergic receptor P2Y, G-protein coupled, 6
Gene Aliases: P2RY6; P2Y6; PP2891
UniProt ID: (Human) Q15077
Entrez Gene ID: (Human) 5031
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.