Invitrogen
Manufacturer Code:PA534093
Catalog # PIPA534093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from 2nd cytoplasmic domain of human P2RY6 / P2Y6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-coupled nucleotide receptor; P2 purinoceptor; P2RY6 G-coupled nucleotide receptor p2 purinoceptor p2y3 receptor p2Y purinoceptor 6 Purinergic receptor p2y6 p2RY6 p2y3 and chick p2Y6 receptor p2Y6 Purinoceptor p2y6; P2Y purinoceptor 6; P2Y6; P2Y6 receptor; pyrimidinergic receptor P2Y, G-protein coupled, 6
Gene Aliases: P2RY6; P2Y6; PP2891
UniProt ID: (Human) Q15077
Entrez Gene ID: (Human) 5031
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.