Invitrogen
Manufacturer Code:PA5112121
Catalog # PIPA5112121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the center region of the Human P2RY6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010204J23Rik G-coupled nucleotide receptor MGC15335 P2 purinoceptor P2ry6 P2Y ATP receptor 6 P2Y purinoceptor 6 p2y3 and chick p2y3 receptor P2Y6 P2Y6 receptor PP2891 Purinoceptor p2y6 pyrimidinergic receptor P2Y G-protein coupled 6 pyrimidinergic receptor P2Y6; G-coupled nucleotide receptor; P2 purinoceptor; P2Y purinoceptor 6; P2Y6; P2Y6 receptor; pyrimidinergic receptor P2Y, G-protein coupled, 6
Gene Aliases: P2RY6; P2Y6; PP2891
UniProt ID: (Human) Q15077
Entrez Gene ID: (Human) 5031
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.