Novus Biologicals
Manufacturer Code:NBP295193100UL
Catalog # NB024548
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 286-365 of human P2RY4 (NP_002556.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: P2P; P2P P2Y4P2Y purinoceptor 4 pyrimidinergic receptor P2Y G-protein coupled 4 UNRNRUUridine nucleotide receptor; P2Y purinoceptor 4; P2Y4; pyrimidinergic receptor P2Y, G-protein coupled, 4; UNR; Uridine nucleotide receptor
Gene Aliases: NRU; P2P; P2RY4; P2Y4; UNR
UniProt ID: (Human) P51582
Entrez Gene ID: (Human) 5030
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.