Invitrogen
Manufacturer Code:MA531822
Catalog # PIMA531822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B4C11 |
Immunogen | Purified recombit fragment of human P2RY4 expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: NRU P2P UNR P2Y4; P2P; P2Y purinoceptor 4; P2Y4; pyrimidinergic receptor P2Y, G-protein coupled, 4; UNR; Uridine nucleotide receptor
Gene Aliases: NRU; P2P; P2RY4; P2Y4; UNR
UniProt ID: (Human) P51582
Entrez Gene ID: (Human) 5030
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.