Invitrogen
Manufacturer Code:PA5102683
Catalog # PIPA5102683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human P2RY4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NRU P2P P2ry4 P2Y ATP receptor 4 P2Y purinoceptor 4 P2y4 P2Y4 receptor P2Y4R P2YR4 purinergic receptor P2Y G-protein coupled 4 purinergic receptor P2Y G-protein coupled 4 purinergic receptor P2Y G-protein coupled 4 pyrimidinergic receptor P2Y G-protein coupled 4 pyrimidinergic receptor P2Y4 RP13-26D14.5 UNR Uridine nucleotide receptor; P2P; P2Y ATP receptor 4; P2Y purinoceptor 4; P2Y4; P2Y4 receptor; purinergic receptor P2Y G-protein coupled 4; purinergic receptor P2Y, G-protein coupled 4; purinergic receptor P2Y, G-protein coupled, 4; pyrimidinergic receptor P2Y, G-protein coupled, 4; UNR; Uridine nucleotide receptor
Gene Aliases: NRU; P2P; P2RY4; P2Y4; P2Y4R; UNR
UniProt ID: (Human) P51582, (Rat) O35811, (Mouse) Q9JJS7
Entrez Gene ID: (Human) 5030, (Rat) 63843, (Mouse) 57385
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.