Novus Biologicals
Manufacturer Code:NBP29475120UL
Catalog # NB024022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 278-377 of human P2RY2 (NP_788086.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP receptor; ATP receptor HP2U MGC20088 MGC40010 P2RU1 P2U nucleotide receptor P2U purinoceptor 1 P2U1 P2UP2UR P2Y purinoceptor 2 P2Y2P2Y2R Purinergic receptor purinergic receptor P2Y G-protein coupled 2 purinergic receptor P2Y2 purinoceptor P2Y2; P2U nucleotide receptor; P2U purinoceptor 1; P2U receptor 1; P2U1; P2Y purinoceptor 2; P2Y2; Purinergic receptor; purinergic receptor P2Y, G-protein coupled, 2; purinoceptor P2Y2
Gene Aliases: HP2U; P2RU1; P2RY2; P2U; P2U1; P2UR; P2Y2; P2Y2R
UniProt ID: (Human) P41231
Entrez Gene ID: (Human) 5029
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.