Novus Biologicals
Manufacturer Code:NBP261753
Catalog # NBP2-61753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5A3F1 |
Immunogen | Purified recombinant fragment of human P2RY14 (AA: extra mix) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: G protein coupled receptor for UDP-glucose; G protein coupled receptor for UDP-glucose GPR105 G-protein coupled receptor 105 KIAA0001G protein-coupled receptor 105 P2Y purinoceptor 14 P2Y(14) receptor P2Y14 P2Y14 receptor purinergic receptor P2Y G-protein coupled 14 UDP-glucose receptor; G protein-coupled receptor 105; G-protein coupled receptor 105; P2Y purinoceptor 14; P2Y(14) receptor; P2Y14; P2Y14 receptor; purinergic receptor P2Y, G-protein coupled, 14; UDP-glucose receptor
Gene Aliases: BPR105; GPR105; KIAA0001; P2RY14; P2Y14
UniProt ID: (Human) Q15391
Entrez Gene ID: (Human) 9934
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.