Invitrogen
Manufacturer Code:MA531817
Catalog # PIMA531817
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2H1G9 |
Immunogen | Purified recombit fragment of human P2RY13 (AA: 1-49) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 86; G-protein coupled receptor 86; G-protein coupled receptor 94; GPCR1 GPR86 GPR94 P2Y13 SP174 FKSG77; P2Y purinoceptor 13; P2Y13; purinergic receptor P2Y G-protein coupled 13; purinergic receptor P2Y, G-protein coupled, 13
Gene Aliases: FKSG77; GPCR1; GPR86; GPR94; P2RY13; P2Y13; SP174
UniProt ID: (Human) Q9BPV8
Entrez Gene ID: (Human) 53829
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.