Novus Biologicals
Manufacturer Code:NBP219660
Catalog # NBP219660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human P2Y12. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-glucose receptor; ADPG-R; ADPG-R Gi-coupled ADP receptor HORK3 HORK3P2Y12 platelet ADP receptor P2T(AC) P2Y purinoceptor 12 P2Y(AC) P2Y(ADP) P2Y(cyc) P2Y12ADP-glucose receptor purinergic receptor P2RY12 purinergic receptor P2Y G-protein coupled 12 putative G-protein coupled receptor SP1999G-protein coupled receptor SP1999; G-protein coupled receptor SP1999; Gi-coupled ADP receptor HORK3; P2T(AC); P2Y purinoceptor 12; P2Y(AC); P2Y(ADP); P2Y(cyc); P2Y12; P2Y12 platelet ADP receptor; purinergic receptor P2RY12; purinergic receptor P2Y, G-protein coupled, 12; putative G-protein coupled receptor; SP1999
Gene Aliases: ADPG-R; BDPLT8; HORK3; P2RY12; P2T(AC); P2Y(12)R; P2Y(AC); P2Y(ADP); P2Y(cyc); P2Y12; SP1999
UniProt ID: (Human) Q9H244
Entrez Gene ID: (Human) 64805
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.