R&D Systems
Manufacturer Code:MAB9305100
Catalog # MAB9305100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 505214 |
Immunogen | NS0 mouse myeloma cell line transfected with human P2Y11/P2RY11 Accession # Q96G91 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: P2RY11 P2Y11P2Y purinoceptor 11 purinergic receptor P2Y G-protein coupled 11 purinergic receptor P2Y11; P2Y purinoceptor 11; P2Y11; purinergic receptor P2Y, G-protein coupled, 11
Gene Aliases: P2RY11; P2Y11
UniProt ID: (Human) Q96G91
Entrez Gene ID: (Human) 5032
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.